Learn More
MSC Starrett Rectangular Gage Block, Steel, 2 in., Grade A1, 1 ea
Catalog No. 5000514934
Encompass
This rectangular steel gage block is designed for precise calibration, layout, and machine setup applications. Engineered for high accuracy and durability, it provides a reliable standard for dimensional measurement.
  • Manufactured from high-quality steel
  • Rectangular shape for versatile positioning
  • Grade A1 accuracy for precise measurements
  • Size of 2 in.
  • Made in the United States
Catalog No. 50-005-14934 Supplier MSC Supplier No. 08470577
This rectangular steel gage block is designed for precise calibration, layout, and machine setup applications. Engineered for high accuracy and durability, it provides a reliable standard for dimensional measurement.
  • Manufactured from high-quality steel
  • Rectangular shape for versatile positioning
  • Grade A1 accuracy for precise measurements
  • Size of 2 in.
  • Made in the United States