Learn More
MSC Starrett Rectangular Gage Block, Steel, Grade A1, 0.108 in., 1 ea.
Catalog No. 5000512342
Encompass
This rectangular gage block is designed for precise measurement and calibration applications in machining environments. Constructed from high-grade steel, it ensures durability and dimensional stability for accurate reference standards.
  • Manufactured from high-grade steel for long-term wear resistance
  • Rectangular shape for versatile handling and inspection
  • Designed for high-precision calibration and measurement tasks
  • Size: 0.108 in.
Catalog No. 50-005-12342 Supplier MSC Supplier No. 98275316
This rectangular gage block is designed for precise measurement and calibration applications in machining environments. Constructed from high-grade steel, it ensures durability and dimensional stability for accurate reference standards.
  • Manufactured from high-grade steel for long-term wear resistance
  • Rectangular shape for versatile handling and inspection
  • Designed for high-precision calibration and measurement tasks
  • Size: 0.108 in.