This rectangular steel gage block is an ultra-precision reference standard. Each block has an engraved identification number for traceability. Made from high-grade alloy steel, it offers exceptional stability, accuracy, and wear resistance. It's heat-treated, hardened, and stabilized through freezing and heating cycles. A final lapping process ensures maximum accuracy and a mirror-like finish. Its rectangular design allows easy wringing to create custom lengths or stacks.
- Provides highly accurate measurements.
- Engraved identification number for traceability.
- High-grade alloy steel provides stability, accuracy, and wear resistance.
- Stabilizing process ensures long-term accuracy.
- Final lapping results in a mirror-like finish.
- Rectangular shape allows for easy wringing to form custom lengths or stacks.