Learn More
MSC Starrett Micrometer Rod, 5/32 in. Diameter, 2.0000 in. Length, 1 ea.
Catalog No. 5000511679
Encompass
This micrometer rod is designed for use with 443 and 445 series micrometer depth gages, providing a reliable extension for precise dimensional measurements.
  • Rod diameter of 5/32 in.
  • Maximum measurement capacity of 2.0000 in.
  • Constructed for integration with depth gage systems
Catalog No. 50-005-11679 Supplier MSC Supplier No. 86417847
This micrometer rod is designed for use with 443 and 445 series micrometer depth gages, providing a reliable extension for precise dimensional measurements.
  • Rod diameter of 5/32 in.
  • Maximum measurement capacity of 2.0000 in.
  • Constructed for integration with depth gage systems