Learn More
MSC Starrett Rubber Dust Guard, For AGD Indicators, 1 inch Range
Catalog No. 5000534494
Encompass
This rubber dust guard is designed to protect the rack of AGD Indicators from foreign matter, especially in adverse gaging conditions. It is compatible with indicators with up to a 1 inch (25 mm) range.
  • Protects indicator rack from foreign matter
  • Suitable for adverse gaging conditions
  • Compatible with AGD Indicators
  • Fits indicators with up to 1 inch (25 mm) range
Catalog No. 50-005-34494 Supplier MSC Supplier No. 86429487
This rubber dust guard is designed to protect the rack of AGD Indicators from foreign matter, especially in adverse gaging conditions. It is compatible with indicators with up to a 1 inch (25 mm) range.
  • Protects indicator rack from foreign matter
  • Suitable for adverse gaging conditions
  • Compatible with AGD Indicators
  • Fits indicators with up to 1 inch (25 mm) range