Learn More
MSC Starrett Screw Pitch Gage, Metric, 0.5-11.5 mm, 28-Leaf, Steel
Catalog No. 5000527382
Encompass
This screw pitch gage is designed for accurately measuring metric screw threads. It features a robust steel construction with 28 leaves, providing a comprehensive range for various pitch sizes from 0.50 to 11.50 mm. The gage is suitable for both internal and external thread location, ensuring precise measurements in a wide array of applications, and includes a 60° thread angle.
  • Metric measurement type
  • Pitch range of 0.50-11.50 mm
  • 28 leaves for diverse thread sizes
  • Constructed from durable steel
  • 60° thread angle for accurate measurement
  • Locking mechanism for secure measurements
  • Suitable for internal and external thread location
Catalog No. 50-005-27382 Supplier MSC Supplier No. 86405933
This screw pitch gage is designed for accurately measuring metric screw threads. It features a robust steel construction with 28 leaves, providing a comprehensive range for various pitch sizes from 0.50 to 11.50 mm. The gage is suitable for both internal and external thread location, ensuring precise measurements in a wide array of applications, and includes a 60° thread angle.
  • Metric measurement type
  • Pitch range of 0.50-11.50 mm
  • 28 leaves for diverse thread sizes
  • Constructed from durable steel
  • 60° thread angle for accurate measurement
  • Locking mechanism for secure measurements
  • Suitable for internal and external thread location