Learn More
MSC Starrett Handle, For 124 Series Inside Micrometers, 6-1/2 in. L, 1 ea.
Catalog No. 5000515239
Encompass
This handle is a precision-engineered component designed for use with 124 and 14M series solid rod inside micrometers. It securely screws into the micrometer head in place of the standard dummy screw to provide added stability and reach.
  • Provides 6.5 in. of length to assist with measurements in deep holes
  • Designed as a replacement or accessory for compatible solid rod inside micrometers
  • Threads directly into the micrometer head for a stable, integrated connection
Catalog No. 50-005-15239 Supplier MSC Supplier No. 86405560
This handle is a precision-engineered component designed for use with 124 and 14M series solid rod inside micrometers. It securely screws into the micrometer head in place of the standard dummy screw to provide added stability and reach.
  • Provides 6.5 in. of length to assist with measurements in deep holes
  • Designed as a replacement or accessory for compatible solid rod inside micrometers
  • Threads directly into the micrometer head for a stable, integrated connection