Learn More
MSC Starrett Rectangular Steel Gage Block, RS 10, AS1 Grade, 10/Set
Catalog No. 5000515652
Encompass
This set of 10 rectangular steel gage blocks is designed for the calibration of micrometers and other precision hand tools. These Grade AS1 blocks provide a reliable reference for verifying measurement accuracy in industrial and laboratory settings.
  • Includes 10 rectangular blocks
  • Grade AS1 precision
  • Constructed from steel
  • Set sizes include 0.105, 0.210, 0.315, 0.420, 0.500, 0.605, 0.710, 0.815, 0.920, and 1.000 in.
  • Made in the USA
Catalog No. 50-005-15652 Supplier MSC Supplier No. 80719537
This set of 10 rectangular steel gage blocks is designed for the calibration of micrometers and other precision hand tools. These Grade AS1 blocks provide a reliable reference for verifying measurement accuracy in industrial and laboratory settings.
  • Includes 10 rectangular blocks
  • Grade AS1 precision
  • Constructed from steel
  • Set sizes include 0.105, 0.210, 0.315, 0.420, 0.500, 0.605, 0.710, 0.815, 0.920, and 1.000 in.
  • Made in the USA