Learn More
MSC Starrett Steel Parallel Set, 6 in, Special Grade Tool Steel, 4 Pairs
Catalog No. 5000528772
Encompass
This steel parallel set is indispensable for inspection, layout work, and various setups on drill presses, milling and grinding machines, and shapers. Made from a special grade of tool steel, the parallels are hardened and accurately ground on all four sides. This set includes four pairs of parallels, each 6 inches in length, and comes in a protective case.
  • Indispensable for inspection and layout work or for various setups.
  • Hardened and accurately ground on four sides.
  • Made from a special grade of tool steel.
  • Furnished in pairs.
  • Six inch length.
  • Set of four pairs.
  • Comes in a protective case.
Catalog No. 50-005-28772 Supplier MSC Supplier No. 58700154
This steel parallel set is indispensable for inspection, layout work, and various setups on drill presses, milling and grinding machines, and shapers. Made from a special grade of tool steel, the parallels are hardened and accurately ground on all four sides. This set includes four pairs of parallels, each 6 inches in length, and comes in a protective case.
  • Indispensable for inspection and layout work or for various setups.
  • Hardened and accurately ground on four sides.
  • Made from a special grade of tool steel.
  • Furnished in pairs.
  • Six inch length.
  • Set of four pairs.
  • Comes in a protective case.