Learn More
MSC Starrett Steel Parallel, Tool Steel, 6 in, 3/4 in, 3/8 in
Catalog No. 5000535088
Encompass
These steel parallels are essential for inspection and layout work, as well as for various setups on machinery. Crafted from a special grade of tool steel, they are hardened and accurately ground to close limits on all four sides. They are furnished in pairs, providing reliable precision in tool rooms and machine shops.
  • Made from a special grade of tool steel, hardened and accurately ground on four sides.
  • Supplied in pairs for convenience.
  • Essential for inspection and layout work.
  • Ideal for setups on drill presses, milling and grinding machines, and shapers.
  • Features a regular steel finish for quick reading.
  • Maintains squareness of 0.0005 in per 1 in.
Catalog No. 50-005-35088 Supplier MSC Supplier No. 58700055
These steel parallels are essential for inspection and layout work, as well as for various setups on machinery. Crafted from a special grade of tool steel, they are hardened and accurately ground to close limits on all four sides. They are furnished in pairs, providing reliable precision in tool rooms and machine shops.
  • Made from a special grade of tool steel, hardened and accurately ground on four sides.
  • Supplied in pairs for convenience.
  • Essential for inspection and layout work.
  • Ideal for setups on drill presses, milling and grinding machines, and shapers.
  • Features a regular steel finish for quick reading.
  • Maintains squareness of 0.0005 in per 1 in.