Learn More
MSC Starrett Steel Square, 4-1/2 in Blade, 3-1/2 in Base
Catalog No. 5000527573
Encompass
This steel square offers precise measurements for various applications. It features a 4-1/2 in blade and a 3-1/2 in base, constructed from high-quality tool steel for durability and accuracy.
  • Accurate to 0.0025 mm
  • Made of high-quality tool steel
  • Meets California Proposition 65 standards
Catalog No. 50-005-27573 Supplier MSC Supplier No. 86401726
Add to Cart
Add to Cart
This steel square offers precise measurements for various applications. It features a 4-1/2 in blade and a 3-1/2 in base, constructed from high-quality tool steel for durability and accuracy.
  • Accurate to 0.0025 mm
  • Made of high-quality tool steel
  • Meets California Proposition 65 standards