Learn More
MSC Starrett Taper Gage, 0.1 to 0.5 Inch Measurement, 8 Leaf, 0.001 Inch Graduation
Catalog No. 5000530233
Encompass
This taper gage is two and a half inches long, made of tempered steel, and features a 0.001 inch graduation. It is designed for accurately determining hole sizes in dies and other work.
  • Made from tempered steel.
  • Features a locking device.
  • Provides 0.001 inch graduation.
  • Has a two and a half inch length.
  • Suitable for determining hole sizes in dies and other work.
Catalog No. 50-005-30233 Supplier MSC Supplier No. 86412335
This taper gage is two and a half inches long, made of tempered steel, and features a 0.001 inch graduation. It is designed for accurately determining hole sizes in dies and other work.
  • Made from tempered steel.
  • Features a locking device.
  • Provides 0.001 inch graduation.
  • Has a two and a half inch length.
  • Suitable for determining hole sizes in dies and other work.