Learn More
MSC Starrett Telescoping Gage, 3-1/2-6 in Range, 3-1/4 in Handle
Catalog No. 5000535036
Encompass
This telescoping gage is used for accurately determining the true size of holes, slots, and recesses up to 6 in (150 mm). The tool must be slightly rocked in the hole being measured to ensure accuracy before it is locked and withdrawn. The final hole size is then obtained by measuring over the gage contacts with a micrometer.
  • Precision ground radius on each gage provides two-point contact for maximum accuracy.
  • Handle, one rigid contact arm, and one spring-tensioned telescoping contact arm.
  • Ends of both contacts are hardened and ground to a radius to allow proper clearance on the smallest hole the gage will enter.
  • Handles can be individually ordered or in larger sizes such as 8 in, 12 in or longer.
Catalog No. 50-005-35036 Supplier MSC Supplier No. 58698259
This telescoping gage is used for accurately determining the true size of holes, slots, and recesses up to 6 in (150 mm). The tool must be slightly rocked in the hole being measured to ensure accuracy before it is locked and withdrawn. The final hole size is then obtained by measuring over the gage contacts with a micrometer.
  • Precision ground radius on each gage provides two-point contact for maximum accuracy.
  • Handle, one rigid contact arm, and one spring-tensioned telescoping contact arm.
  • Ends of both contacts are hardened and ground to a radius to allow proper clearance on the smallest hole the gage will enter.
  • Handles can be individually ordered or in larger sizes such as 8 in, 12 in or longer.