Learn More
MSC Starrett Test Indicator Kit, 0.001 in Graduation, 18 in Max, 0-20-0 Dial
Catalog No. 5000534539
Encompass
This test indicator kit is a crankshaft distortion dial gage designed for precise measurements. It features a 0.001 in graduation and an 18 in maximum measurement, suitable for various applications requiring high accuracy. The kit includes a balancing attachment and is equipped with a non-breakable crystal and movable bezel for reliable performance.
  • Includes 12 pieces for comprehensive use.
  • Designed as a crankshaft distortion dial gage.
  • Provides 0.001 in graduation for detailed readings.
  • Offers a maximum measurement of 18 in.
  • Equipped with a 0-20-0 dial reading.
  • Includes a 2-3/8 to 18 in balancing attachment.
  • Features one extension.
  • Boasts a non-breakable crystal for durability.
  • Comes with a movable bezel for user convenience.
  • Meets California PROP 65 standards.
Catalog No. 50-005-34539 Supplier MSC Supplier No. 86423886
This test indicator kit is a crankshaft distortion dial gage designed for precise measurements. It features a 0.001 in graduation and an 18 in maximum measurement, suitable for various applications requiring high accuracy. The kit includes a balancing attachment and is equipped with a non-breakable crystal and movable bezel for reliable performance.
  • Includes 12 pieces for comprehensive use.
  • Designed as a crankshaft distortion dial gage.
  • Provides 0.001 in graduation for detailed readings.
  • Offers a maximum measurement of 18 in.
  • Equipped with a 0-20-0 dial reading.
  • Includes a 2-3/8 to 18 in balancing attachment.
  • Features one extension.
  • Boasts a non-breakable crystal for durability.
  • Comes with a movable bezel for user convenience.
  • Meets California PROP 65 standards.