Learn More
MSC Starrett Test Indicator Kit, 3 Pc, 0.0300" Max, 0-15-0
Catalog No. 5000516203
Encompass
This 3-piece Test Indicator Kit offers precision measurement for various applications. It features a maximum measurement of 0.0300" and a dial reading of 0-15-0, with a fine graduation of 0.000500" and a 0.120" ball. The kit includes a dial test indicator, a contact point, and a body clamp, designed for reliable use without a shank in its case.
  • Accuracy
  • Hard chrome-plated point
  • Swiveling tubular body
  • Universal friction holder
  • Jeweled lever action
  • Positive reversing switch
  • Interchangeable contact point
  • Includes dial test indicator, contact point, body clamp
  • Complies with California Prop 65 standards
Catalog No. 50-005-16203 Supplier MSC Supplier No. 86424272
This 3-piece Test Indicator Kit offers precision measurement for various applications. It features a maximum measurement of 0.0300" and a dial reading of 0-15-0, with a fine graduation of 0.000500" and a 0.120" ball. The kit includes a dial test indicator, a contact point, and a body clamp, designed for reliable use without a shank in its case.
  • Accuracy
  • Hard chrome-plated point
  • Swiveling tubular body
  • Universal friction holder
  • Jeweled lever action
  • Positive reversing switch
  • Interchangeable contact point
  • Includes dial test indicator, contact point, body clamp
  • Complies with California Prop 65 standards