This thickness gauge features tempered steel feeler stock in a convenient rewindable dispenser roll. It is designed for precise measurement and adjustment tasks, making it ideal for various applications such as adjusting tappets, spark plugs, checking bearing clearances, and shimming in fixturing and die work.
- Housed in convenient rewindable dispenser rolls.
- Useful for cutting off the required length for adjusting tappets, spark plugs, distributor points, checking bearing clearances and gear play, fitting pistons, rings and pins, and gaging narrow slots.
- Also useful for shimming in fixturing and die work.
- Marked every 6 in or 150 mm with a line for easy, exact dispensing.
- Compact, sturdy plastic rewindable dispenser case.
- Case provides the ability to snip off the desired length without any waste.