Learn More
MSC Starrett Universal Back Plunger Dial Indicator, 5 mm, 0.01 mm, 0-100 Dial
Catalog No. 5000531579
Encompass
This universal back plunger dial indicator is designed for a wide range of measurement tasks. It features precise graduations and a robust construction, ensuring accurate and reliable readings. The indicator is easy to operate, allowing for quick setup and measurement of variations in workpieces.
  • Versatile for a wide range of applications.
  • Accurate and reliable for precise measurements.
  • Simple to operate with smooth motion.
  • Rugged construction ensures durability.
  • Includes three hardened contact points for adaptable use.
  • Features a 6.3 mm (1/4 in) shank diameter.
  • Offers a 5 mm measurement range.
  • Provides 0.01 mm graduations.
  • Equipped with a 0-100 dial reading.
Catalog No. 50-005-31579 Supplier MSC Supplier No. 86407384
This universal back plunger dial indicator is designed for a wide range of measurement tasks. It features precise graduations and a robust construction, ensuring accurate and reliable readings. The indicator is easy to operate, allowing for quick setup and measurement of variations in workpieces.
  • Versatile for a wide range of applications.
  • Accurate and reliable for precise measurements.
  • Simple to operate with smooth motion.
  • Rugged construction ensures durability.
  • Includes three hardened contact points for adaptable use.
  • Features a 6.3 mm (1/4 in) shank diameter.
  • Offers a 5 mm measurement range.
  • Provides 0.01 mm graduations.
  • Equipped with a 0-100 dial reading.