Learn More
MSC Starrett Vernier Caliper, 72 in., 0.001 in. Graduation, Tool Steel, 1 ea.
Catalog No. 5000511802
Encompass
This vernier caliper is a high-precision measuring instrument designed for versatility and durability in demanding industrial applications. Featuring a 72 in. (182.88 cm) range and 0.001 in. graduations, it utilizes an open-face design with long 50-division vernier scales to ensure accurate and easy readings. The tool is engineered for structural rigidity, featuring hardened, ground, and lapped measuring surfaces for long-term consistency.
  • Satin chrome finish eliminates glare for improved visibility
  • Fine adjustment screw-type nut enables precise measurements
  • Locking nut securely holds measurements in place
  • Hardened tool steel construction for enhanced durability
  • Adjustable slide provides a longer bearing surface for squareness
  • Machine-divided graduations ensure accuracy
Catalog No. 50-005-11802 Supplier MSC Supplier No. 86405271
This vernier caliper is a high-precision measuring instrument designed for versatility and durability in demanding industrial applications. Featuring a 72 in. (182.88 cm) range and 0.001 in. graduations, it utilizes an open-face design with long 50-division vernier scales to ensure accurate and easy readings. The tool is engineered for structural rigidity, featuring hardened, ground, and lapped measuring surfaces for long-term consistency.
  • Satin chrome finish eliminates glare for improved visibility
  • Fine adjustment screw-type nut enables precise measurements
  • Locking nut securely holds measurements in place
  • Hardened tool steel construction for enhanced durability
  • Adjustable slide provides a longer bearing surface for squareness
  • Machine-divided graduations ensure accuracy