Learn More
MSC Starrett Vernier Height Gage, 0-12 in, 0.001 in, Satin Chrome
Catalog No. 5000531591
Encompass
The Starrett 255Z-12 Vernier Height Gage is a precise measuring instrument designed for general-purpose applications. It features a 0 to 12 inch measuring range with 0.001 inch graduations and a durable satin chrome finish. The height gage comes with a protective case, providing convenience and longevity.
  • 50 division vernier scales readable to 0.001 or 0.02 mm without magnification.
  • Flush fitting vernier scales eliminate parallax.
  • Sharp, easy-to-read black lines against a satin chrome finish.
  • Scriber and base designed for direct reading from zero.
  • Quick adjustment slide with screw-type adjusting nut for precise positioning.
  • Hardened, stabilized bar and balanced base design prevent vibration.
  • Hardened, ground, and lapped base for sure grip and easy movement.
  • Scribes lines and measures with dial indicators or electronic probes.
Catalog No. 50-005-31591 Supplier MSC Supplier No. 86411600
The Starrett 255Z-12 Vernier Height Gage is a precise measuring instrument designed for general-purpose applications. It features a 0 to 12 inch measuring range with 0.001 inch graduations and a durable satin chrome finish. The height gage comes with a protective case, providing convenience and longevity.
  • 50 division vernier scales readable to 0.001 or 0.02 mm without magnification.
  • Flush fitting vernier scales eliminate parallax.
  • Sharp, easy-to-read black lines against a satin chrome finish.
  • Scriber and base designed for direct reading from zero.
  • Quick adjustment slide with screw-type adjusting nut for precise positioning.
  • Hardened, stabilized bar and balanced base design prevent vibration.
  • Hardened, ground, and lapped base for sure grip and easy movement.
  • Scribes lines and measures with dial indicators or electronic probes.