Learn More
MSC Starrett Wiggler/Center Finder, Complete with Case, 4 Attachments
Catalog No. 5000511678
Encompass
This wiggler/center finder set is designed for precise alignment and location of working centers. Its interchangeable attachments adapt to numerous applications, allowing for quick and accurate positioning of work with the machine spindle.
  • Attachments snap into chuck without collet nut removal
  • Attachments clamped by ball swivel-joint for angular adjustment or true concentricity
  • Ball contact for locating work
  • Disc contact for use in confined areas
  • Offset indicator holder for use with test indicators
  • Awl point
  • Comes with a protective case
Catalog No. 50-005-11678 Supplier MSC Supplier No. 86425311
This wiggler/center finder set is designed for precise alignment and location of working centers. Its interchangeable attachments adapt to numerous applications, allowing for quick and accurate positioning of work with the machine spindle.
  • Attachments snap into chuck without collet nut removal
  • Attachments clamped by ball swivel-joint for angular adjustment or true concentricity
  • Ball contact for locating work
  • Disc contact for use in confined areas
  • Offset indicator holder for use with test indicators
  • Awl point
  • Comes with a protective case