Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Starswab II™ Collection and Transport Systems: Amies Charcoal Medium

Catalog No. 1490583 Shop All Starplex Scientific Products
Change view
Click to view available options
Shaft Material:
Plastic
Straight Wire
Quantity:
100/Cs.
50 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shaft Material Quantity
14-905-83 Straight Wire 100/Cs.
14-905-68 Plastic 50 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-905-83 Supplier Starplex™ Scientific Supplier No. SP131W
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protective overskirt on cap

  • Designed for collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
  • Swab tips available in single, double or straight wire format
  • Applicators have color-coded caps with overskirt and biohazard plug-cap rings; extra-long swab shafts; and a labeled, round-bottom, spiral transport tube
  • Transport tubes contain 4mL of semi-solid media
  • Spiral media chamber within a seamless polypropylene tube protects sample and keeps it immersed in media during transport
  • Labels preprinted with lot number, expiration date and product code with space reserved for recording patient information
  • Gamma sterilized and wrapped in sterile, easy-to-peel envelope

Amies Charcoal Medium

  • Improves the survival of fastidious organisms (especially Neisseria gonorrhoeae) by neutralizing any toxic components present

Specifications

Product Type Collection and Transport System
Media Type Modified Amies Charcoal Gel
Certifications/Compliance CE
Closure Color Black
For Use With (Application) Collecting, transporting and maintaining the viability of aerobic, anaerobic and facultative microorganisms
Length (English) 6 in.
Length (Metric) 15.2 cm
Packaging Bag
Quantity 100/Cs.
Shaft Material Straight Wire
Storage Requirements 4°C to 25°C
Tip Material Rayon
Tip Type Minitip Flocked
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.