Learn More
Perkin Elmer US LLC Start Up Kit for SPB 50-48
Catalog No. 502103716
Encompass_Preferred
SPB Start Up kits contain all consumable items required to operate the system including: RackLock DigiTUBEs and Caps, disposable Watch Glasses, DigiFILTERs, RackLock Racks and Storage Racks.
Catalog No. 50-210-3716 Supplier Perkin Elmer US LLC Supplier No. N9308002
Add to Cart
Add to Cart
Request a Quote
SPB Start Up kits contain all consumable items required to operate the system including: RackLock DigiTUBEs and Caps, disposable Watch Glasses, DigiFILTERs, RackLock Racks and Storage Racks.