Learn More
World Precision Instrument Start-up Kit for TBM4M
SDP
Catalog No. 502529300
Encompass
Get up and running with your TBM4M Transbridge Amplifier
The TBM4M is a transbridge, a four-channel analog transducer manifold, that is specifically designed to amplify the output voltage signal from transducers of pressure, force, displacement, temperature and a wide variety of other signal sources.
Catalog No. 50-252-9300 Supplier World Precision Instrument Supplier No. 3718
Get up and running with your TBM4M Transbridge Amplifier
The TBM4M is a transbridge, a four-channel analog transducer manifold, that is specifically designed to amplify the output voltage signal from transducers of pressure, force, displacement, temperature and a wide variety of other signal sources.