Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Start-up Solution, Reversed Phase, UV Detection, Liquid

Catalog No. 50201708 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This start-up solution contains Uracil, ethyl, and propyl Parabens, designed for use in liquid chromatography (LC) systems for installation purposes. It has a 12-month shelf life and requires storage at room temperature. This product is classified as hazardous material, which necessitates special freight handling and may incur additional charges.

  • Contains Uracil, ethyl, and propyl Parabens.
  • Designed for liquid chromatography (LC) systems.
  • Separation mode: Reversed Phase.
  • Detection method: UV.
  • Formulation: Liquid.
  • Storage condition: Room Temperature.
  • Shelf life: 12 months.
  • Shipping condition: Ambient.
  • Type: Installation.
  • Hazardous material.

Catalog No. 50-201-708 Supplier Waters Corp Supplier No. WAT034544
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This start-up solution contains Uracil, ethyl, and propyl Parabens, designed for use in liquid chromatography (LC) systems for installation purposes. It has a 12-month shelf life and requires storage at room temperature. This product is classified as hazardous material, which necessitates special freight handling and may incur additional charges.

  • Contains Uracil, ethyl, and propyl Parabens.
  • Designed for liquid chromatography (LC) systems.
  • Separation mode: Reversed Phase.
  • Detection method: UV.
  • Formulation: Liquid.
  • Storage condition: Room Temperature.
  • Shelf life: 12 months.
  • Shipping condition: Ambient.
  • Type: Installation.
  • Hazardous material.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.