Learn More
Electron Microscopy Sciences K-kits Starter Box
SDP
Catalog No. 501928049
Encompass_Preferred
The Starter Box contains all of the essentials for K-kit loading. It consists of glues, a beaker, four stirring sticks, and two stainless steel thin needles. Used with the K-kit for TEM observation of liquid samples.
Catalog No. 50-192-8049 Supplier Electron Microscopy Sciences Supplier No. K7268
The Starter Box contains all of the essentials for K-kit loading. It consists of glues, a beaker, four stirring sticks, and two stainless steel thin needles. Used with the K-kit for TEM observation of liquid samples.