Learn More
OHAUS™ Starter™ Conductivity Electrode
Catalog No. 01922385 Shop All OHAUS Products
Change view
Click to view available options
Measurement Ranges:
0.055 to 300 μS/cm
50 to 2000 μS/cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Ranges
01-922-385 0.055 to 300 μS/cm
01-922-384 50 to 2000 μS/cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-922-385 Supplier OHAUS™ Supplier No. 30681117
Add to Cart
Add to Cart
The Starter™ Conductivity Electrode features a glass shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation for accurate measurements.
  • Designed for accurate and precise measurement
  • Can be used with both bench and portable meters
  • Temperature sensor features automatic temperature compensation
TRUSTED_SUSTAINABILITY
Specifications
Cell Constant 0.1 cm ±0.02
Type Conductivity Electrode
Connector Types Mini-DIN
Material Glass
Measurement Ranges 0.055 to 300 μS/cm