Learn More
OHAUS™ Starter™ Conductivity Electrode STCON8 with Flow Chamber
Catalog No. 01922386 Shop All OHAUS Products
Change view
Click to view available options
Connector Types:
Mini-DIN
Catalog No. Connector Types
01-922-386 Mini-DIN
1 options
Catalog No. 01-922-386 Supplier OHAUS™ Supplier No. 30681235
Add to Cart
Add to Cart
The Starter™ Conductivity Electrode STCON8 with Flow Chamber features a glass shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation for accurate measurements. Flow chamber produces accurate measurements.
  • Designed for accurate and precise measurement
  • Can be used with both bench and portable meters
  • Flow chamber optimizes the sample flow volume and creates mixing patterns that produce accurate and stable measurements
TRUSTED_SUSTAINABILITY
Specifications
Cell Constant 0.1 cm ±0.02
Type Conductivity Electrode
Connector Types Mini-DIN
Material Glass
Measurement Ranges 0.055 to 300 μS/cm