Learn More
Braintree Scientific Starter Kit for OEM Syringe Pump, for BS-500
SDP
Catalog No. 501955428
Encompass
Allows user to interactively communicate to the pump with a computer Includes: power supply, Pump-to-PC network cable (CBL-PC-PUMP-7), and user manual **Sold for research purposes only and not for human use**
Catalog No. 50-195-5428 Supplier Braintree Scientific Supplier No. OEMKIT120V
Allows user to interactively communicate to the pump with a computer Includes: power supply, Pump-to-PC network cable (CBL-PC-PUMP-7), and user manual **Sold for research purposes only and not for human use**