Learn More
Electron Microscopy Sciences Tripod Polisher Starter Kit for Tripod Polisher
SDP
Catalog No. 50980885
Encompass
Diamond Lapping Film (0.5, 1, 3, 6, 15, and 30 back); Multitex Polishing Cloth; 8" Glass Plate; 0.05 um Colloidal Silica; Mounting Wax; Squeegee; Slotted TEM Grids, Cleaner; Swabs; Petri Dish, Filter Paper, Tweezers.
Catalog No. 50-980-885 Supplier Electron Microscopy Sciences Supplier No. 5011805
Diamond Lapping Film (0.5, 1, 3, 6, 15, and 30 back); Multitex Polishing Cloth; 8" Glass Plate; 0.05 um Colloidal Silica; Mounting Wax; Squeegee; Slotted TEM Grids, Cleaner; Swabs; Petri Dish, Filter Paper, Tweezers.