Learn More
USA Scientific Inc Starter Pack, Con Tubes 25 mL
Catalog No. 502745281
Encompass
Eppendorf Conical Tubes 25 mL with screw cap, Starter Pack, PCR clean, 200 Tubes, 4 Single Tube Stands, 6 adapters for rotors with bore for 50 mL conical tubes
Catalog No. 50-274-5281 Supplier USA Scientific Inc Supplier No. 40112500
Eppendorf Conical Tubes 25 mL with screw cap, Starter Pack, PCR clean, 200 Tubes, 4 Single Tube Stands, 6 adapters for rotors with bore for 50 mL conical tubes