Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Starter Pak No. 1
GREENER_CHOICE

Catalog No. 215366 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-536-6 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-536-6 Supplier Gibco™ Supplier No. 215366
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Features a combination of yeast- and soy-based peptones.

  • Three of the products in this pack have been ultra-filtered to reduce endotoxin levels
  • The yeast products add a mixture of peptides, amino acids, carbohydrates (simple and complex), nucleosides, and vitamins to any medium formulation
  • Starter Pak No. 1 provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for use in human and animal health applications and contains:
    • Difco TC Yeastolate UF and Bacto TC Yeastolate—these peptones are ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins
    • Difco Yeast Extract UF and Bacto Yeast Extract, Technical—these peptones support optimal growth of many microbial species for a variety of human and animal health vaccines
    • Difco Phytone Supplement, UF—this enzymatic digest of soy is an excellent, nutritious source of carbohydrates and is used in mammalian cell culture. This peptone works well alone and when blended with yeast-based peptones
  • Our Starter Paks provide conveniently packaged samples of the peptones most commonly used in mammalian cell culture and microbial fermentation bioproduction processes. Starter Paks are tailored for specific applications, including the production of monoclonal antibodies, recombinant proteins, and vaccines.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Culture Type Microbial
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Human and animal health applications
Quantity 1 Ea.
Source Animal origin-free, yeast and soy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.