Learn More
Description
- It provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for human and animal vaccine production and contains:
- Bacto Yeast Extract and Gibco Yeast Extract—these peptones contain a mixture of peptides, amino acids, carbohydrates, and vitamins to support optimal growth in microbial species, and are ideal for the production of vaccines
- Bacto TC Yeastolate—this peptone is ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins, as well as vaccine applications
- Phytone Peptone and Difco Soytone—these soy-based supplements provide a nutritious source of carbohydrates for successful use in microbial fermentation. Blending these soy peptones with yeast peptones has been shown to provide additional benefit to cultures.
- Bacto Malt Extract—a water-soluble portion of malted barley, this peptone also provides carbohydrates for a variety of microbial fermentation processes
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Human and animal vaccine productions |
| Quantity | 1 Ea. |
| Source | Animal origin-free, yeast and soy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
