Learn More
OHAUS™ Starter™ pH Electrode
SCI_ED
Catalog No. p-7236596 Shop All OHAUS Products
Change view
Click to view available options
Junction:
Ceramic
Fiber Pin
Material:
Glass
Plastic
pH Range:
0 to 14
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Junction Material pH Range
S05310 Ceramic Glass 0 to 14
S05309 Fiber Pin Plastic 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05310 Supplier OHAUS™ Supplier No. 30656037/EMD
Add to Cart
Add to Cart
The Starter™ pH Electrode features a durable shaft built to withstand daily use. Includes a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
  • Designed for accurate and precise measurement
  • Can be used with both bench and portable meters
  • Temperature sensor features automatic temperature compensation
TRUSTED_SUSTAINABILITY
Specifications
Connector Types BNC
Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 5°C to 90°C
Type pH Electrode