Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Starter™ pH Meter ST2200

Catalog No. 01922379 Shop All OHAUS Products
Change view
Click to view available options
Type:
pH Meter
pH Meter with Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-922-379 pH Meter
01-922-378 pH Meter with Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-922-379 Supplier OHAUS™ Supplier No. 30656033
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Starter™ pH Meter ST2200 is an affordable benchtop pH Meter that clearly displays information with five distinctly marked keys, allowing users to operate with minimal training.

  • Equipped with a large 3.5 in. LCD display
  • Features a 99 item memory for measurements and last calibration
  • Features auto-buffer recognition and auto-endpoint modes that make measurement and calibration simple
  • Choose between just the ST2200 meter with electrode holder and a full kit including ST2200 meter, electrode holder, ST320 electrode, 4 pH buffer mini kits (50 mL)
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1.5
Accuracy (pH) ±0.01
Data Management Details Datalog for Measurements and Last Calibration
Includes ST2200 Meter, Electrode Holder
Measurement Modes Auto-stop, Continuous
Calibration Linear
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Calibration Points 1 to 3
Temperature Range (Metric) 0.0°C to 100.0°C
Display Type LCD
Backlight Option Yes
Data Points 99 Sets
Log Function Type Automatic Data Logging
Type pH Meter
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.