Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand2Mind Starter Science Graduated Cylinders
SCI_ED

Catalog No. S02766
Change view
Click to view available options
Class Size:
1 Student
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02766 1 Student
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02766 Supplier Hand2Mind Supplier No. 95813
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Graduated cylinders make it easy for students to measure and pour liquids. Set includes 5 graduated cylinders (10, 25, 50, 100, and 250mL). Bases keep the cylinders stable, and colored rings slide to make measuring accurate.

  • Children feel like real scientists as they use our graduated cylinders to measure and pour liquids.
  • Made from clear, durable plastic, have nonslip bases, and include protective collars.

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Student
Product Type Graduated Cylinder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.