Learn More
Description
- Encourage hands-on experimentation and observation while introducing Science/STEM activities at home or in the classroom.
- Sturdy test tubes feature a flat bottom for stability.
- Set includes a convenient storage rack so children can easily carry their experiments from place to place and a reusable cleaning brush to make cleanup a breeze.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | Test Tubes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
