Learn More
Macherey-Nagel Starter Set A
Supplier:  Macherey-Nagel 740682
Encompass_Preferred
Starter Set A for processing NucleoSpin 8-well strips under vacuum on the NucleoVac 96 Vacuum Manifold or similar manifolds - 2 Column Holders A, 12 NucleoSpin Dummy Strips
Catalog No. NC1229311