Learn More
Hach Company Starter set for Lico
Supplier:  Hach Company LZM354
Encompass
Starter Kit consists of ADDISTA color standard color solutionsGlass round cuvettes 11 mm and PMMA Rectangular cuvettes 50 mm
Catalog No. 50-002-07340