Learn More
Cell Signaling Technology Stat Antibody Sampler Kit, Western Blot, 1 Kit
SDP
Catalog No. 502051320
Encompass
The Stat Antibody Sampler Kit provides an economical solution for investigating the expression of multiple Stat proteins, specifically Stat1, Stat3, Stat5, and Stat6. Each kit includes a comprehensive set of primary and secondary antibodies required to conduct two Western blot experiments.
  • Includes primary and secondary antibodies for Western blotting
  • Allows for the simultaneous examination of Stat1, Stat3, Stat5, and Stat6
  • Provides sufficient materials for two Western blot experiments
  • Features high-quality monoclonal and polyclonal antibodies
  • Optimized for research applications in immunology and Jak/Stat signaling
Catalog No. 50-205-1320 Supplier Cell Signaling Technology Supplier No. 9939T
The Stat Antibody Sampler Kit provides an economical solution for investigating the expression of multiple Stat proteins, specifically Stat1, Stat3, Stat5, and Stat6. Each kit includes a comprehensive set of primary and secondary antibodies required to conduct two Western blot experiments.
  • Includes primary and secondary antibodies for Western blotting
  • Allows for the simultaneous examination of Stat1, Stat3, Stat5, and Stat6
  • Provides sufficient materials for two Western blot experiments
  • Features high-quality monoclonal and polyclonal antibodies
  • Optimized for research applications in immunology and Jak/Stat signaling