Learn More
Amstat Industries Staticmaster 1U200 1" Ionizers
SDP
Catalog No. NC9900754
Encompass
The 1U200 1" Ionizer is a self-powered static eliminator. Initial activity level is 250 microcuries. Its small profile makes it an ideal solution for in-balance elimination. This small profile ionizer offers "zero-volts balance" technology, which means that it can effectively remove all of the static charge present in your media and samples. This ensures accurate weighing every time.
Catalog No. NC9900754 Supplier Amstat Industries Supplier No. 1U200
The 1U200 1" Ionizer is a self-powered static eliminator. Initial activity level is 250 microcuries. Its small profile makes it an ideal solution for in-balance elimination. This small profile ionizer offers "zero-volts balance" technology, which means that it can effectively remove all of the static charge present in your media and samples. This ensures accurate weighing every time.