Learn More
ECM Bioscience Stat Phospho-Regulation Antibody Sampler Kit, ECM Biosciences
Catalog No. 502068831
Encompass_Preferred
The Stat phospho-regulation antibody sampler kit can be used to examine the level of phosphorylation of Stat1 and Stat5, as well as the total expression level of Stat1, Stat3, and Stat5. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms
Catalog No. 50-206-8831 Supplier ECM Bioscience Supplier No. SK6030
The Stat phospho-regulation antibody sampler kit can be used to examine the level of phosphorylation of Stat1 and Stat5, as well as the total expression level of Stat1, Stat3, and Stat5. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms