Learn More
EKF Diagnostics STAT-Site™ Blood Glucose β-Ketone Strips
SDP
Catalog No. 22022652 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Packaging Type:
Box
Catalog No. Packaging Type
22-022-652 Box
1 options
This item is not returnable. View return policy
Catalog No. 22-022-652 Supplier EKF Diagnostics Supplier No. 7020050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Blood Glucose Control Solution that provides result in 10 seconds
  • 1 μL sample size
  • Uses capillary or venous whole blood
Specifications
Type Test Strip
Packaging Type Box