Learn More
EKF Diagnostics STAT-Site™ Blood Glucose Control Solution
SDP
Catalog No. 22022650 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Type:
Control Solution
Catalog No. Type
22-022-650 Control Solution
1 options
This item is not returnable. View return policy
Catalog No. 22-022-650 Supplier EKF Diagnostics Supplier No. 7014204
Add to Cart
Add to Cart
This item is not returnable. View return policy
Blood Glucose Control Solution includes one W2 (norm) and one B3 (high).
Contains 1 x W2 (norm) and 1 x B3 (high)
  • Includes: 1 x W2 (norm) and 1 x B3 (high)
Specifications
Type Control Solution