Learn More
Stanbio™ Accessories for STAT-Site™ M B-HB Meter (B-Hydroxybutyrate System)
Shop All Stanbio Laboratory Products
Description
- Quantitative testing–results in two minutes
- Metabolic status, insulin uptake, and end of catabolism
- STAT differential diagnosis between lacticacidosis and ketoacidosis
- Blood ketone testing for diagnosis and monitoring of ketoacidosis
- Tests for Β-hydroxybutyrate, the most accurate marker for measuring Diabetic Ketoacidosis (DBA)
- Monitors the status of ketoacidosis, documents insulin uptake and defines the end of catabolism
- An alternative to qualitative nitroprusside tests
- Virtually no interference from captopril and other sulfhydryl drugs
- Can also aid in STAT differential diagnosis between lacticacidosis and ketoacidosis
- Use with Stanbio Stat-Site™ Analyzer for convenient, bedside testing
Specifications
Specifications
| Type | Diluent (strip/card) |
| For Use With (Application) | Blood Ketone testing |
| Includes | STAT-Site M BHB Diluent |
| Quantity | 6 x 3 mL |