Learn More
Stanbio™ Accessories for STAT-Site™ M B-HB Meter (B-Hydroxybutyrate System)
SDP
Catalog No. SB303500 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Type:
BHB Controls (strip/card)
BHB Linearity (strip/card)
Diluent (strip/card)
Quantity:
1 x 1 mL
2 x 3 mL
6 x 3 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
SB303500 BHB Linearity (strip/card) 1 x 1 mL
SB202000 Diluent (strip/card) 6 x 3 mL
SB303000 BHB Controls (strip/card) 2 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SB303500 Supplier Stanbio™ Supplier No. 303500
Assurance from false-positives, producing more confidence in results and greater test utility
  • Quantitative testing–results in two minutes
  • Metabolic status, insulin uptake, and end of catabolism
  • STAT differential diagnosis between lacticacidosis and ketoacidosis
  • Blood ketone testing for diagnosis and monitoring of ketoacidosis
  • Tests for Β-hydroxybutyrate, the most accurate marker for measuring Diabetic Ketoacidosis (DBA)
  • Monitors the status of ketoacidosis, documents insulin uptake and defines the end of catabolism
  • An alternative to qualitative nitroprusside tests
  • Virtually no interference from captopril and other sulfhydryl drugs
  • Can also aid in STAT differential diagnosis between lacticacidosis and ketoacidosis
  • Use with Stanbio Stat-Site™ Analyzer for convenient, bedside testing
Specifications
Type BHB Linearity (strip/card)
For Use With (Application) Blood Ketone testing
Includes STAT-Site M BHB Linearity
Quantity 1 x 1 mL