Learn More
BIOCHEMICAL DIAGNOSTICS INC Stat-Skreen17, Pos & Neg, 2X10Ml Doa Control.
Catalog No. NC1808125
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
DRUGS OF ABUSE 17 drug (300ng Opiate Cutoff) Urine Control Postive and Negative 20ml kitbox (1X10ml each level). Includes THC50, COC300, PCP25, OPI300,AMP1000, MAMP1000,BARB300,BZO300, MTD300,PPX300, TCA1000, OXY100, MDMA500, BUP10, EDDP300, FYL25
Catalog No. NC1808125 Supplier BIOCHEMICAL DIAGNOSTICS INC Supplier No. 702003
This item has a minimum qty of 2 per supplier requirements.
DRUGS OF ABUSE 17 drug (300ng Opiate Cutoff) Urine Control Postive and Negative 20ml kitbox (1X10ml each level). Includes THC50, COC300, PCP25, OPI300,AMP1000, MAMP1000,BARB300,BZO300, MTD300,PPX300, TCA1000, OXY100, MDMA500, BUP10, EDDP300, FYL25