Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ζ-Stat trisodium | 31894-34-5 | 97.0% | 450.31 g·mol⁻¹ | C10H5Na3O10S3 | 100MG
SDP

Catalog No. 5000238378
Encompass_Preferred

ζ-Stat trisodium is the trisodium salt of NSC37044 and a selective, atypical protein kinase C zeta (PKC-ζ) inhibitor reported to have an IC50 of approximately 5 μM. It has demonstrated in vitro antitumor activity, including reduced proliferation and induced apoptosis in melanoma cell lines. This research chemical is supplied as a solid with reported purity of 97.0%.

  • Selective PKC-ζ inhibitor; IC50 ≈ 5 μM.
  • Demonstrated reduction of melanoma cell proliferation in vitro.
  • Reported to induce apoptosis in cancer cell lines.
  • Provided as a light yellow solid for research use.
  • High purity: 97.0%.
  • Molecular weight 450.31 g·mol⁻¹; formula C10H5Na3O10S3.
  • Available in 100 mg research quantities.

Catalog No. 50-002-38378 Supplier Medchemexpress LLC Supplier No. HY123979A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ζ-Stat trisodium is the trisodium salt of NSC37044 and a selective, atypical protein kinase C zeta (PKC-ζ) inhibitor reported to have an IC50 of approximately 5 μM. It has demonstrated in vitro antitumor activity, including reduced proliferation and induced apoptosis in melanoma cell lines. This research chemical is supplied as a solid with reported purity of 97.0%.

  • Selective PKC-ζ inhibitor; IC50 ≈ 5 μM.
  • Demonstrated reduction of melanoma cell proliferation in vitro.
  • Reported to induce apoptosis in cancer cell lines.
  • Provided as a light yellow solid for research use.
  • High purity: 97.0%.
  • Molecular weight 450.31 g·mol⁻¹; formula C10H5Na3O10S3.
  • Available in 100 mg research quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.