Learn More
Medchemexpress LLC ζ-Stat trisodium | 31894-34-5 | 97.0% | 450.31 g/mol | C10H5Na3O10S3 | 5MG
SDP
Catalog No. 5000235046
Encompass_Preferred
ζ-Stat trisodium (trisodium;8-hydroxynaphthalene-1,3,6-trisulfonate) is the trisodium salt of NSC37044 and a selective inhibitor of atypical protein kinase C zeta (PKC-ζ). It is provided as a light yellow to light brown solid for biochemical and cellular research and has been reported to reduce melanoma cell proliferation and induce apoptosis in vitro.
Catalog No. 50-002-35046 Supplier Medchemexpress LLC Supplier No. HY123979A5MG
ζ-Stat trisodium (trisodium;8-hydroxynaphthalene-1,3,6-trisulfonate) is the trisodium salt of NSC37044 and a selective inhibitor of atypical protein kinase C zeta (PKC-ζ). It is provided as a light yellow to light brown solid for biochemical and cellular research and has been reported to reduce melanoma cell proliferation and induce apoptosis in vitro.