Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ζ-Stat trisodium (NSC37044 trisodium) | 31894-34-5 | 97.0% | 450.31 | C10H5Na3O10S3 | 50MG
SDP

Catalog No. 5000236933
Encompass_Preferred

ζ-Stat trisodium is a small-molecule inhibitor of protein kinase C zeta (PKC-ζ) used in biochemical and cellular research. It is supplied as a light yellow to light brown solid with a reported purity of 97.0%, molecular formula C10H5Na3O10S3, and molecular weight 450.31 g/mol. The compound is typically provided in small, preweighed quantities suitable for laboratory studies.

  • Inhibits protein kinase C zeta (PKC-ζ) activity.
  • CAS number 31894-34-5.
  • Purity 97.0%.
  • Molecular formula C10H5Na3O10S3; molecular weight 450.31 g/mol.
  • Solid, light yellow to light brown appearance.
  • Available in multiple small research quantities (mg scale).

Catalog No. 50-002-36933 Supplier Medchemexpress LLC Supplier No. HY123979A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ζ-Stat trisodium is a small-molecule inhibitor of protein kinase C zeta (PKC-ζ) used in biochemical and cellular research. It is supplied as a light yellow to light brown solid with a reported purity of 97.0%, molecular formula C10H5Na3O10S3, and molecular weight 450.31 g/mol. The compound is typically provided in small, preweighed quantities suitable for laboratory studies.

  • Inhibits protein kinase C zeta (PKC-ζ) activity.
  • CAS number 31894-34-5.
  • Purity 97.0%.
  • Molecular formula C10H5Na3O10S3; molecular weight 450.31 g/mol.
  • Solid, light yellow to light brown appearance.
  • Available in multiple small research quantities (mg scale).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.