Learn More
Aviva Systems Biology Corp STAT1 Antibody - N-terminal region (ARP36979_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36979_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against STAT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse STAT1. Alternate names: AA408197, 2010005J02RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-56302